1-Bromoperfluorohexane 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Perfluorohexyl bromide
PC6085W-100G
PC6085W-25G
77432-480EA
381.04
USD
77432-480
77432-482
1-Bromoperfluorohexane 98%
1-Bromoperfluorohexane
Formula:
BrF₂C(CF₂)₄CF₃ MW: 398.95 g/mol Boiling Pt: 97…101 °C Density: 1.871 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042349 CAS Number: 335-56-8 EINECS: 206-391-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...