2-Chloro-5-(trifluoromethyl)benzoic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

PC5584-5G
77431-734EA 21.26 USD
77431-734
2-Chloro-5-(trifluoromethyl)benzoic acid 96%
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR