Methyl perfluorohexadecanoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC5138-5G
77431-150EA 68.18 USD
77431-150
Methyl perfluorohexadecanoate 95%
Methyl perfluorohexadecanoate
Formula: C₁₇H₃F₃₁O₂
Boiling Pt: 167 °C/50 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00236617
CAS Number: 165457-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR