2'-Fluoro-4'-methoxyacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3817B-100G PC3817B-25G PC3817B-5G
77429-016EA 108 USD
77429-016 77429-018 77429-020
2'-Fluoro-4'-methoxyacetophenone 98%
2'-Fluoro-4'-methoxyacetophenone

Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).

Formula: C₉H₉FO₂
Boiling Pt: 80…85 °C/0.1 mmHg
Melting Pt: 51…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042290
CAS Number: 74457-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR