5-Fluoro-3-pyridineboronic acid 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC3624-1G PC3624-25G PC3624-5G
77428-888EA 28.52 USD
77428-888 77428-890 77428-892
5-Fluoro-3-pyridineboronic acid 95+%
5-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR