1,2-Difluoro-4-nitrobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4-Difluoronitrobenzene

PC2862-100G PC2862-500G
77427-938EA 21.4 USD
77427-938 77427-940
1,2-Difluoro-4-nitrobenzene 99%
1,2-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80
Density: 1.437 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR