3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC28261-250MG
77427-870EA 39.74 USD
77427-870
3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
3-Bromo-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₂BrClF₃N₃
MW: 300.463 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406873
CAS Number: 1823182-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR