2-Chloro-3-fluoro-5-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC1401-1G
77427-036EA 21.26 USD
77427-036
2-Chloro-3-fluoro-5-methylpyridine
2-Chloro-3-fluoro-5-methylpyridine
Formula: C₆H₅ClFN
Storage Temperature: Ambient
MDL Number: MFCD06658238
CAS Number: 34552-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR