2-Bromo-4,5-difluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene

PC1393N-5G
77427-014EA 28.52 USD
77427-014
2-Bromo-4,5-difluorophenol 98%
2-Bromo-4,5-difluorophenol
Formula: C₆H₃BrF₂O
Density: 1.829 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00070751
CAS Number: 166281-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR