1-Bromo-3-chloro-5-fluorobenzene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3-Chloro-5-fluorobromobenzene
PC1367M-100G
PC1367M-25G
PC1367M-5G
77426-976EA
54
USD
77426-976
77426-978
77426-980
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene
Formula:
C₆H₃BrClF Density: 1.72 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00070747 CAS Number: 33863-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...