4-Amino-3-(trifluoromethoxy)benzoic acid 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC1100BD-1G
PC1100BD-25G
PC1100BD-5G
77426-786EA
21.26
USD
77426-786
77426-788
77426-790
4-Amino-3-(trifluoromethoxy)benzoic acid 97%
4-Amino-3-(trifluoromethoxy)benzoic acid
Formula:
C₈H₆F₃NO₃ Melting Pt: 168…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00205143 CAS Number: 175278-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...