2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC10142-500MG
77426-538EA 131.42 USD
77426-538
2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid
2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid
Formula: C₁₃H₅F₆N₃O₂
Melting Pt: 281…283 °C
Storage Temperature: Ambient
MDL Number: MFCD03425718
CAS Number: 439094-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR