Pentafluorophenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid

PC0869-5G PC0869-10G PC0869-1G
77426-416EA 59.1 USD
77426-416 77426-412 77426-414
Pentafluorophenylboronic acid 97%
Pentafluorophenylboronic acid
Formula: C₆H₂BF₅O₂
Melting Pt: 250…255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR