3-(Trifluoromethoxy)-DL-phenylglycine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC0517-1G
PC0517-250MG
77426-260EA
182.36
USD
77426-260
77426-262
3-(Trifluoromethoxy)-DL-phenylglycine
3-(Trifluoromethoxy)-DL-phenylglycine
Formula:
C₉H₈F₃NO₃ MW: 235.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631460 CAS Number: 261952-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...