3,5-Difluoro-o-phenylenediamine

Supplier: APOLLO SCIENTIFIC
Warning

PC0143-1G PC0143-25G PC0143-5G
77426-070EA 21.4 USD
77426-070 77426-072 77426-074
3,5-Difluoro-o-phenylenediamine
3,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
Melting Pt: 48.5…49.5 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00973899
CAS Number: 2369-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR