3-Chloro-2-fluorobenzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC0060-1G PC0060-5G
77426-052EA 28.26 USD
77426-052 77426-054
3-Chloro-2-fluorobenzyl alcohol 97%
3-Chloro-2-fluorobenzyl alcohol
Formula: C₇H₆ClFO
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD01631549
CAS Number: 261723-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR