Ethyl benzoate 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: Benzoic acid ethyl ester

OR925081-100G
77425-156EA 59.1 USD
77425-156
Ethyl benzoate 95%
Ethyl benzoate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR