5-Bromo-1H-indazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR9243-5G OR9243-1G OR9243-25G
77425-136EA 63.16 USD
77425-136 77425-126 77425-128
5-Bromo-1H-indazole 97%
5-Bromo-1H-indazole
Formula: C₇H₅BrN₂
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00839493
CAS Number: 53857-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR