Methyl 3-(1-aminoethyl)benzoate hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR918899-250MG
77424-994EA 88 USD
77424-994
Methyl 3-(1-aminoethyl)benzoate hydrochloride 95%
Methyl 3-(1-aminoethyl)benzoate hydrochloride
Formula: C₁₀H₁₃NO₂·ClH
MW: 215.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432494
CAS Number: 1263378-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR