3-Aminobutan-1-ol 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Amino-butan-1-ol

OR912855-10G OR912855-1G OR912855-5G
77424-918EA 66.22 USD
77424-918 77424-920 77424-922
3-Aminobutan-1-ol 97%
3-Aminobutan-1-ol
Formula: C₄H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD03412694
CAS Number: 2867-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR