Pirfenidone 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

OR911941-1G OR911941-5G
77424-876EA 22.42 USD
77424-876 77424-878
Pirfenidone 98%
Pirfenidone
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00866047
CAS Number: 53179-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR