2-Methylpyrimidine-4-carbaldehyde 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR904478-1G
OR904478-250MG
OR904478-5G
77424-768EA
183.38
USD
77424-768
77424-770
77424-772
2-Methylpyrimidine-4-carbaldehyde 98%
2-Methylpyrimidine-4-carbaldehyde
Formula:
C₆H₆N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD09832936 CAS Number: 1004-17-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...