5-Bromoisoquinoline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR8906-100G OR8906-25G OR8906-5G
77424-678EA 207.84 USD
77424-678 77424-680 77424-682
5-Bromoisoquinoline 98%
5-Bromoisoquinoline
Formula: C₉H₆BrN
Boiling Pt: 95…97 °C/0.1 mmHg
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD01646405
CAS Number: 34784-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR