5-Bromoquinoline 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR8604-1G OR8604-25G OR8604-5G
77424-566EA 21.26 USD
77424-566 77424-568 77424-570
5-Bromoquinoline 97%
5-Bromoquinoline
Formula: C₉H₆BrN
Boiling Pt: 105…107 °C/1 mmHg
Melting Pt: 41…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00234481
CAS Number: 4964-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR