5-Bromo-2-hydroxypyrimidine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR8599-1G
OR8599-25G
OR8599-5G
77424-554EA
21.26
USD
77424-554
77424-556
77424-558
5-Bromo-2-hydroxypyrimidine
5-Bromo-2-hydroxypyrimidine
Formula:
C₄H₃BrN₂O Storage Temperature: Ambient |
MDL Number:
MFCD01318956 CAS Number: 38353-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...