5-Bromo-2-(dimethylamino)pyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-N,N-dimethylpyridin-2-amine

OR7730-1G
77424-306EA 21.26 USD
77424-306
5-Bromo-2-(dimethylamino)pyridine
5-Bromo-2-(dimethylamino)pyridine
Formula: C₇H₉BrN₂
MW: 201.0600 g/mol
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00099486
CAS Number: 26163-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR