4-(Methylsulfonyl)-2-nitrobenzaldehyde 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7647-250MG OR7647-1G
77424-256EA 41.78 USD
77424-256 77424-254
4-(Methylsulfonyl)-2-nitrobenzaldehyde 95+%
4-(Methylsulfonyl)-2-nitrobenzaldehyde
Formula: C₈H₇NO₅S
Storage Temperature: Ambient
MDL Number: MFCD04037929
CAS Number: 849035-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR