4-Hexylphenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR7224-1G OR7224-250MG OR7224-5G
77424-132EA 28.52 USD
77424-132 77424-134 77424-136
4-Hexylphenylboronic acid 97%
4-Hexylphenylboronic acid
Formula: C₁₂H₁₉BO₂
MDL Number: MFCD04039029
CAS Number: 105365-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR