Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR62216-1G
77423-710EA 544.04 USD
77423-710
Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%
Methyl 4-bromo-2-iodo-5-methoxybenzoate
Formula: C₉H₈BrIO₃
MW: 370.97 g/mol
CAS Number: 2384857-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR