Methyl-3,4,5-trihydroxybenzoate 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

OR5903-100G
77423-264EA 50.94 USD
77423-264
Methyl-3,4,5-trihydroxybenzoate 98%
Methyl 3,4,5-trihydroxybenzoate
Formula: C₈H₈O₅
MW: 184.15 g/mol
Boiling Pt: 450
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00002194
CAS Number: 99-24-1
Merck Index: 13,04366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR