6-Chlorouracil 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione

OR5447-25G
77422-892EA 21.4 USD
77422-892
6-Chlorouracil 98%
6-Chlorouracil
Formula: C₄H₃ClN₂O₂
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014595
CAS Number: 4270-27-3
EINECS: 224-258-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR