Potassium thiocyanate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5319-500G OR5319-100G OR5319-25G
77422-816EA 247.58 USD
77422-816 77422-812 77422-814
Potassium thiocyanate 98%
Potassium thiocyanate
Formula: KSCN
Boiling Pt: 500 °C
Melting Pt: 172…175 °C
Density: 1.89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR