Potassium thiocyanate 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Potassium rhodanide
OR5319-500G
OR5319-100G
OR5319-25G
77422-816EA
247.58
USD
77422-816
77422-812
77422-814
Potassium thiocyanate 98%
Potassium thiocyanate
Formula:
KSCN Boiling Pt: 500 °C Melting Pt: 172…175 °C Density: 1.89 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...