Sulfolane 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR51759-100G
77422-496EA 28.52 USD
77422-496
Sulfolane 99%
Sulpholane
Formula: C₄H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR