Ethyl 3-Amino-4-hydroxybenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR471083-10G OR471083-1G OR471083-5G
77421-930EA 533.86 USD
77421-930 77421-932 77421-934
Ethyl 3-Amino-4-hydroxybenzoate
Ethyl 3-Amino-4-hydroxybenzoate
Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD09961411
CAS Number: 13052-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR