2-Iodophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4682-100G OR4682-25G OR4682-500G
77421-896EA 63.16 USD
77421-896 77421-898 77421-900
2-Iodophenol 98%
2-Iodophenol
Formula: C₆H₅IO
Boiling Pt: 186…187 °C/160 mmHg
Melting Pt: 37…43 °C
Density: 1.947 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR