2-Iodophenol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR4682-100G
OR4682-25G
OR4682-500G
77421-896EA
63.16
USD
77421-896
77421-898
77421-900
2-Iodophenol 98%
2-Iodophenol
Formula:
C₆H₅IO Boiling Pt: 186…187 °C/160 mmHg Melting Pt: 37…43 °C Density: 1.947 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013963 CAS Number: 533-58-4 EINECS: 208-569-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...