6-Iodo-o-toluic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR46539-1G OR46539-250MG OR46539-5G
77421-844EA 116.14 USD
77421-844 77421-846 77421-848
6-Iodo-o-toluic acid 95%
6-Iodo-o-toluic acid
Formula: C₈H₇IO₂
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01310823
CAS Number: 54811-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR