2,6-Dichloro-3-cyanopyrazine

Supplier: APOLLO SCIENTIFIC
Warning

OR45109-10G OR45109-5G
77421-582EA 188.48 USD
77421-582 77421-584
2,6-Dichloro-3-cyanopyrazine
2,6-Dichloro-3-cyanopyrazine
Formula: C₅HCl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD13193460
CAS Number: 313339-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR