Sodium methanesulfinate, technical

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methanesulphinic acid, sodium salt, Methanesulfinic acid, sodium salt

OR42101-25G
77421-338EA 21.32 USD
77421-338
Sodium methanesulfinate, technical
Sodium methanesulphinate
Formula: CH₃NaO₂S
MW: 102.09 g/mol
Melting Pt: 222…226 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040392
CAS Number: 20277-69-4
EINECS: 243-669-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR