2-Cyclopropyl-4-iodoaniline

Supplier: APOLLO SCIENTIFIC
Warning

OR42059-250MG OR42059-1G
77421-336EA 48.9 USD
77421-336 77421-334
2-Cyclopropyl-4-iodoaniline
2-Cyclopropyl-4-iodoaniline
Formula: C₉H₁₀IN
Storage Temperature: Freezer
MDL Number: MFCD27938271
CAS Number: 1704065-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR