Methyl 4-azaindole-3-carboxylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate, 3-(Methoxycarbonyl)-1H-pyrrolo[3,2-b]pyridine

OR41124-50MG
77421-274EA 20.34 USD
77421-274
Methyl 4-azaindole-3-carboxylate 97%
Methyl 4-azaindole-3-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864655
CAS Number: 952800-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR