5-Bromo-2,7-dimethyl-2H-indazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR40141-250MG
77421-012EA 41.78 USD
77421-012
5-Bromo-2,7-dimethyl-2H-indazole 95%
5-Bromo-2,7-dimethyl-2H-indazole
Formula: C₉H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028637
CAS Number: 1146637-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR