5-Acetylthiophene-2-carboxaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Acetyl-5-formylthiophene, 5-Acetyl-2-thenaldehyde, 5-Acetylthiophene-2-carbaldehyde, 2-Ethanoyl-5-formylthiophene

OR4009-100MG
77420-956EA 25.8 USD
77420-956 77420-962 77420-964
5-Acetylthiophene-2-carboxaldehyde
5-Acetylthiophene-2-carboxaldehyde
Formula: C₇H₆O₂S
MW: 154.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00509280
CAS Number: 4565-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR