2,4-Dichloro-5-nitropyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR3891-100G OR3891-25G
77420-778EA 158.94 USD
77420-778 77420-780
2,4-Dichloro-5-nitropyrimidine
2,4-Dichloro-5-nitropyrimidine
Formula: C₄HCl₂N₃O₂
Boiling Pt: 0.9 °C/0.5 mmHg
Melting Pt: 28…32 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00127867
CAS Number: 49845-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR