3-Methyl-isothiazol-4-ylamine

Supplier: APOLLO SCIENTIFIC
Warning

OR346368-1G OR346368-250MG OR346368-50MG
77420-360EA 588.88 USD
77420-360 77420-362 77420-364
3-Methyl-isothiazol-4-ylamine
3-Methyl-isothiazol-4-ylamine
Formula: C₄H₆N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD10700204
CAS Number: 53483-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR