8-Acetyl-7-hydroxy-4-methylcoumarin

Supplier: APOLLO SCIENTIFIC
Warning

OR345062-5G
77420-296EA 358.44 USD
77420-296
8-Acetyl-7-hydroxy-4-methylcoumarin
Formula: C₁₂H₁₀O₄
Melting Pt: 170 °C
Storage Temperature: Ambient
CAS Number: 2555-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR