4-(Dimethylamino)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR345014-25G OR345014-5G
77420-292EA 58.08 USD
77420-292 77420-294
4-(Dimethylamino)benzonitrile 98%
4-(Dimethylamino)benzonitrile
Formula: C₉H₁₀N₂
Boiling Pt: 318 °C
Melting Pt: 70…76 °C
MDL Number: MFCD00001815
CAS Number: 1197-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR