4-(1-Pyrrolylsulfonyl)benzeneboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR3432-100MG OR3432-1G OR3432-500MG
77420-278EA 110.04 USD
77420-278 77420-280 77420-282
4-(1-Pyrrolylsulfonyl)benzeneboronic acid 98%
4-(1-Pyrrolylsulfonyl)benzeneboronic acid
Formula: C₁₀H₁₀BNO₄S
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD08689522
CAS Number: 913835-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR