3-Bromo-4-fluoroaniline 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC9908-100G
PC9908-25G
77418-766EA
41.78
USD
77418-766
77418-768
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 124 °C/11 mmHg Melting Pt: 29…36 °C Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788559 CAS Number: 656-64-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...